Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554847 Supplier Invitrogen™ Supplier No. PA554847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CDFCLDDTRR VKAVKSCLTC MVNYCEEHLQ PHQVNIKLQS HLLTEPVKDH NWRYCPAHHS PLSAFCCPDQ QCICQDCCQE HSGHTIVSLD AARRDKEAEL QCTQLDLERK LKLNENAISR LQANQKSVLV SVSEVKAVAE MQFGE Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 74%.

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trim16
Gene Accession No. O95361
Gene Alias 9130006M08Rik; AI482483; E3 ubiquitin-protein ligase TRIM16; EBBP; Estrogen-responsive B box protein; TRIM16; tripartite motif containing 16; tripartite motif protein 16; tripartite motif-containing 16; tripartite motif-containing protein 16
Gene Symbols Trim16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM16. Recombinant protein control fragment (Product #RP-89323).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.