Learn More
Invitrogen™ TRIM16 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TRIM16.
Specifications
Specifications
| Antigen | TRIM16 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide; pH 7.4 |
| Gene | Trim16 |
| Gene Accession No. | O95361, Q99PP9 |
| Gene Alias | 9130006M08Rik; AI482483; E3 ubiquitin-protein ligase TRIM16; EBBP; Estrogen-responsive B box protein; TRIM16; tripartite motif containing 16; tripartite motif protein 16; tripartite motif-containing 16; tripartite motif-containing protein 16 |
| Gene Symbols | Trim16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.