Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM16 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5110515

Catalog No. PIPA5110515


Only null left
Add to Cart

Description

Description

Immunogen sequence: MAELDLMAPG PLPRATAQPP APLSPDSGSP SPDSGSASPV EEEDVGSSEK LGRETEEQDS DSAEQGDPAG EGKEVLCDFC LDDTRRVKAV KSCLTCMVNY CEEHLQPHQV NIKLQSHLLT EPVKDHNWRY CPAHHSPLSA FCCPDQQCIC QDCCQEHSGH TIVSLDAARR DKEAELQCTQ LDLERKLKLN ENAISRLQAN QKSVLVSVSE.

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM16
Polyclonal
Unconjugated
Trim16
9130006M08Rik; AI482483; E3 ubiquitin-protein ligase TRIM16; EBBP; Estrogen-responsive B box protein; TRIM16; tripartite motif containing 16; tripartite motif protein 16; tripartite motif-containing 16; tripartite motif-containing protein 16
Rabbit
Affinity Chromatography
RUO
10626, 303214, 94092
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.77 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95361, Q99PP9
Trim16
Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human TRIM16 (NP_006461.3).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.