Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM17 Monoclonal Antibody (2E11)
GREENER_CHOICE

Catalog No. PIMA521862
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521862 Supplier Invitrogen™ Supplier No. MA521862
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LPNRLLTKVA EMAQQHPGLQ KQDLCQEHHE PLKLFCQKDQ SPICVVCRES REHRLHRVLP AEEAVQGYKL KLEEDMEYLR EQITRTGNLQ AREEQSLAEW QGKVKERRER.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM17
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2E11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TRIM17
Gene Accession No. Q9Y577
Gene Alias E3 ubiquitin-protein ligase TRIM17; RBCC; RING finger protein 16; RING finger protein terf; RING-type E3 ubiquitin transferase TRIM17; RNF16; terf; Testis RING finger protein; Trim17; tripartite motif containing 17; tripartite motif protein 17; tripartite motif protein TRIM17; tripartite motif-containing 17; tripartite motif-containing protein 17; Unknown (protein for MGC:134518)
Gene Symbols TRIM17
Host Species Mouse
Immunogen TRIM17 (NP_057186.1, 75 a.a. ~ 184 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51127
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.