Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584223 Supplier Invitrogen™ Supplier No. PA584223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDRQGHSLEL LLLQLEERST QGPLQMLQDM KEPLSRKNNV SVQCPEVAPP TRPRTVCRVP GQIEVLRGFL E Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 65%.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM17
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM17
Gene Accession No. Q9Y577
Gene Alias E3 ubiquitin-protein ligase TRIM17; RBCC; RING finger protein 16; RING finger protein terf; RING-type E3 ubiquitin transferase TRIM17; RNF16; terf; Testis RING finger protein; Trim17; tripartite motif containing 17; tripartite motif protein 17; tripartite motif protein TRIM17; tripartite motif-containing 17; tripartite motif-containing protein 17; Unknown (protein for MGC:134518)
Gene Symbols TRIM17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM17. Recombinant protein control fragment (Product #RP-93606).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.