Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB1001218
Description
TRIM2 Polyclonal specifically detects TRIM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| KIAA0517tripartite motif protein 2, RING finger protein 86, RNF86tripartite motif protein TRIM2, tripartite motif containing 2, tripartite motif-containing 2, tripartite motif-containing protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23321 | |
| Human, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 10 ug/ml, Immunohistochemistry 6 - 12 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 6 - 12 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_056086.2 | |
| TRIM2 | |
| Peptide with sequence DSGNHCFKVYRYLQ corresponding to C-Terminus according to NP_056086.2, NP_001123539.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_056086.2; NP_001123539.1). Cross-reactivity with TRIM3 has been demonstrated. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction