Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM2 Antibody (OTI2B10), DyLight 488, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP274606G

Catalog No. NBP274606G


Only null left
Add to Cart

Description

Description

TRIM2 Monoclonal specifically detects TRIM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

TRIM2
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
TRIM2
Recombinant protein fragment corresponding to amino acids 1-100 and 1500-1600 of human TRIM2 (NP_775105) produced in HEK293T cells.
0.1 mL
23321.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry
OTI2B10
Western Blot, Flow Cytometry
KIAA0517tripartite motif protein 2, RING finger protein 86, RNF86tripartite motif protein TRIM2, tripartite motif containing 2, tripartite motif-containing 2, tripartite motif-containing protein 2
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.