Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM2 Mouse anti-Human, Clone: OTI1D2, lyophilized, TrueMAB™
SDP

Catalog No. 501660564
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-0564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-0564 Supplier OriGene Supplier No. CF501559
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.

Specifications

Antigen TRIM2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1D2
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TRIM2
Gene Accession No. Q9C040
Gene Alias CMT2R, RNF86
Gene Symbols TRIM2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-100 and 645-744 of human TRIM2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23321
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.