Learn More
TRIM2 Mouse anti-Human, Rat, Clone: OTI4E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501551

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.Specifications
TRIM2 | |
Monoclonal | |
Unconjugated | |
TRIM2 | |
CMT2R, RNF86 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23321, 361970 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI4E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
D3ZQG6, Q9C040 | |
Trim2 | |
Human recombit protein fragment corresponding to amino acids 1-100 and 645-744 of human TRIM2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.