Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557430
Description
Immunogen sequence: PMSVTITTKD KDGELCKTGN AYLTAELSTP DGSVADGEIL DNKNGTYEFL YTVQKEGDFT LSLRLYDQHI RGSPFKLKVI RSADVSPT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
Specifications
TRIM2 | |
Polyclonal | |
Unconjugated | |
Trim2 | |
CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23321 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C040 | |
Trim2 | |
Recombinant protein corresponding to Human TRIM2. Recombinant protein control fragment (Product #RP-96625). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction