Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518152
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic filaments. Its function has not been identified.
Specifications
| TRIM2 | |
| Polyclonal | |
| Unconjugated | |
| Trim2 | |
| CMT2R; E3 ubiquitin-protein ligase TRIM2; KIAA0517; mKIAA0517; narf; Neural activity-related RING finger protein; RING finger protein 86; RING-type E3 ubiquitin transferase TRIM2; RNF86; TRIM2; tripartite motif containing 2; tripartite motif protein 2; tripartite motif protein TRIM2; tripartite motif-containing 2; tripartite motif-containing protein 2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 23321, 361970 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| D3ZQG6, Q9C040 | |
| Trim2 | |
| Synthetic peptide sequence (DSGNHCFKVYRYLQ) corresponding to the C-terminus amino acids of TRIM2 (aa 731-744). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction