Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM21 Mouse anti-Human, CoraLite 488, Clone: 1E8B9, Proteintech
SDP

Catalog No. 502257307 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7307 Supplier Proteintech Group Inc Supplier No. CL48867136100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TRIM21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human, mouse, rat antigen. The TRIM21 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.

Specifications

Antigen TRIM21
Applications Immunofluorescence
Classification Monoclonal
Clone 1E8B9
Conjugate CoraLite 488
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6737
Target Species Human, Rat, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.