Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM21/SSA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092648 20 μL
NB092647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092648 Supplier Novus Biologicals Supplier No. NBP30380920UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM21/SSA1 Polyclonal specifically detects TRIM21/SSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen TRIM21/SSA1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81RING finger protein 81, ro(SS-A), RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-330 of human TRIM21/SSA1 (NP_003132.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6737
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.