Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trim22 Antibody (OTI1D11), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274608A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274608A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274608A94 Supplier Novus Biologicals Supplier No. NBP274608AF594
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Trim22 Monoclonal specifically detects Trim22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Trim22
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1D11
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 50 kDa-stimulated trans-acting factor, E3 ubiquitin-protein ligase TRIM22, EC 6.3.2, EC 6.3.2.-, GPSTAF50, RING finger protein 94, RNF94stimulated trans-acting factor (50 kDa), Staf-50, STAF50staf-50, tripartite binding motif 22, tripartite motif containing 22, tripartite motif protein TRIM22, tripartite motif-containing 22, Tripartite motif-containing protein 22
Gene Symbols TRIM22
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 61-406 of human Trim22 (NP_006065) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10346.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.