Learn More
Invitrogen™ TRIM22 Monoclonal Antibody (OTI1D3)

Description
TRIM22 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TRIM22 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TRIM22 |
| Gene Accession No. | Q8IYM9 |
| Gene Alias | 50 kDa-stimulated trans-acting factor; E3 ubiquitin-protein ligase TRIM22; GPSTAF50; RING finger protein 94; RING-type E3 ubiquitin transferase TRIM22; RNF94; STAF50; Staf-50; stimulated trans-acting factor (50 kDa); TRIM22; tripartite binding motif 22; tripartite motif containing 22; tripartite motif protein TRIM22; tripartite motif-containing 22; tripartite motif-containing protein 22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.