Learn More
TRIM22 Mouse anti-Human, Clone: OTI1A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505295

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRIM22 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. It localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects.Specifications
TRIM22 | |
Monoclonal | |
Unconjugated | |
TRIM22 | |
GPSTAF50, RNF94, STAF50 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10346 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IYM9 | |
TRIM22 | |
Human recombit protein fragment corresponding to amino acids 61-406 of human TRIM22 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.