Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551964 Supplier Invitrogen™ Supplier No. PA551964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEVVKECQEK LQVALQRLIK EDQEAEKLED DIRQERTAWK NYIQIERQKI LKGFNEMRVI LDNEEQRELQ KLEEGEVNVL DNLAAATDQL VQQRQDASTL ISDLQRRLRG SSVEMLQDVI DVMKRSESWT LKKPKSVSKK LKSVFRV Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.

TRIM22 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. It localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon's antiviral effects. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon's antiviral effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM22
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM22
Gene Accession No. Q8IYM9
Gene Alias 50 kDa-stimulated trans-acting factor; E3 ubiquitin-protein ligase TRIM22; GPSTAF50; RING finger protein 94; RING-type E3 ubiquitin transferase TRIM22; RNF94; STAF50; Staf-50; stimulated trans-acting factor (50 kDa); TRIM22; tripartite binding motif 22; tripartite motif containing 22; tripartite motif protein TRIM22; tripartite motif-containing 22; tripartite motif-containing protein 22
Gene Symbols TRIM22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM22. Recombinant protein control fragment (Product #RP-102255).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.