Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trim22/Staf-50 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12330R

Description
TRIM22 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. It localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects.Specifications
Trim22/Staf-50 | |
Polyclonal | |
Unconjugated | |
TRIM22 | |
50 kDa-stimulated trans-acting factor; E3 ubiquitin-protein ligase TRIM22; GPSTAF50; RING finger protein 94; RING-type E3 ubiquitin transferase TRIM22; RNF94; STAF50; Staf-50; stimulated trans-acting factor (50 kDa); TRIM22; tripartite binding motif 22; tripartite motif containing 22; tripartite motif protein TRIM22; tripartite motif-containing 22; Tripartite motif-containing protein 22 | |
Rabbit | |
Protein A | |
RUO | |
10346 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8IYM9 | |
TRIM22 | |
KLH conjugated synthetic peptide derived from human Trim22. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction