Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597010 Supplier Invitrogen™ Supplier No. PA597010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATLVVNKLG AGVDSGRQGS RGTAVVKVLE CGVCEDVFSL QGDKVPRLLL CGHTVCHDCL TRLPLHGRAI RCPFDRQVTD LGDSGVWGLK KNFALLELLE RLQNGPIGQY GAAEESIGIS GESIIRCDED EAHLASVYCT VCATHLCSEC SQVTHSTKTL AKHRRVPLAD KPHEKTMCSQ HQVHAIEFVC LEEGCQTSPL MCCVCKEYGK HQGHKHSVLE PEANQIRASI LDMAHCIRTF TEEISDYSRK LVGIVQHIEG GEQIVEDGIG MAHTEHVPGT; Positive Samples: PC-12, Mouse spleen, Mouse brain; Cellular Location: Endomembrane system, Golgi apparatus membrane, Lysosome membrane.

Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM23
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIM23
Gene Accession No. P36406, P36407, Q8BGX0
Gene Alias 6330516O20Rik; ADP-ribosylation factor domain protein 1; ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; AI450195; ARD1; Ard-1; ARF domain protein 1; ARFD1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; RNF46; TRIM23; tripartite motif containing 23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; tripartite motif-containing protein 23
Gene Symbols TRIM23
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM23 (NP_0016471).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373, 81002, 81003
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.