Learn More
Invitrogen™ TRIM23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534624
Description
Recommended positive controls: HeLaS3. Predicted reactivity: Rhesus Monkey (100%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
Specifications
TRIM23 | |
Polyclonal | |
Unconjugated | |
TRIM23 | |
6330516O20Rik; ADP-ribosylation factor domain protein 1; ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; AI450195; ARD1; Ard-1; ARF domain protein 1; ARFD1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; RNF46; TRIM23; tripartite motif containing 23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; tripartite motif-containing protein 23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
373, 81003 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.67 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P36406, Q8BGX0 | |
TRIM23 | |
Synthetic peptide corresponding to a region within amino acids 1 and 44 of TRIM23 (Uniprot ID#P36406). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.