Learn More
TRIM24 Mouse anti-Human, Clone: OTI1A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802796

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
TRIM24 | |
Monoclonal | |
Unconjugated | |
TRIM24 | |
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15164 | |
TRIM24 | |
Human recombit protein fragment corresponding to amino acids 706-1016 of human TRIM24 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.