Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111177 Supplier Invitrogen™ Supplier No. PA5111177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPVGGSYNL PSLPDIDCSS TIMLDNIVRK DTNIDHGQPR PPSNRTVQSP NSSVPSPGLA GPVTMTSVHP PIRSPSA Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 94%.

The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM24
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM24
Gene Accession No. O15164
Gene Alias A130082H20Rik; Ab2-427; AI447469; D430004I05Rik; E3 ubiquitin-protein ligase TRIM24; hTIF1; PTC6; RING finger protein 82; RING-type E3 ubiquitin transferase TIF1-alpha; RNF82; TF1A; Tif1; Tif1a; TIF1alpha; TIF1-alpha; Transcription intermediary factor 1-alpha; transcriptional intermediary factor 1; transcriptional intermediary factor 1, alpha; Trim24; tripartite motif containing 24; tripartite motif-containing 24; tripartite motif-containing protein 24
Gene Symbols TRIM24
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TRIM24. Recombinant protein control fragment (Product #RP-100381).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.