Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588875 Supplier Invitrogen™ Supplier No. PA588875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2, A-431, Mouse uterus; Cellular Location: Cytoplasm Immunogen sequence: SLSQASADLE ATLRHKLTVM YSQINGASRA LDDVRNRQQD VRMTANRKVE QLQQEYTEMK ALLDASETTS TRKIKEEEKR VNSKFDTIYQ ILLKKKSEIQ TLKEEIEQSL TKRDEFEFLE KASKLRGIST KPVYIPEVEL NHKLIKGIHQ STIDLKNELK QCIGRLQEPT PSSGDPGEHD PASTHKSTRP.

TRIM25, also known as estrogen responsive finger protein (EFP) is a member of the RING finger-B box-coiled coil family and is a downstream target of estrogen receptor a. Mice lacking this gene displayed an underdeveloped uterus and reduced estrogen responsiveness demonstrating that this protein is essential for estrogen dependent proliferation. TRIM25 expression is increased in breast carcinoma where it promotes the growth of breast tumor by functioning as an E3 ubiquitin ligase that targets the negative cell cycle checkpoint 14-3-3s. Recent reports have indicated that TRIM25 E3 ubiquitin ligase is essential for the retinoic-acid-inducible gene 1 (RIG-1) signaling pathway to elicit host antiviral innate immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM25
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TRIM25
Gene Accession No. Q14258, Q61510
Gene Alias AA960166; AL022677; E3 ubiquitin/ISG15 ligase TRIM25; Efp; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; RNF147; Trim25; tripartite motif containing 25; tripartite motif protein 25; tripartite motif protein TRIM25; tripartite motif-containing 25; tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Z147; Zfp147; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147; Znf147
Gene Symbols TRIM25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 190-379 of human TRIM25 (NP_0050732).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217069, 7706
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.