Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM25 Rabbit anti-Human, Clone: 4Q0D2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | TRIM25 |
---|---|
Clone | 4Q0D2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TRIM25 Monoclonal antibody specifically detects TRIM25 in Human samples. It is validated for Western BlotSpecifications
TRIM25 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Zinc Finger | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7706 | |
IgG | |
Affinity purified |
4Q0D2 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TRIM25 (Q14258). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title