Learn More
Invitrogen™ TRIM26 Monoclonal Antibody (1G3)

Description
Peptide Sequence: REKILNHLST LRRDRDKIQG FQAKGEADIL AALKKLQDQR QYIVAEFEQG HQFLREREEH LLEQLAKLEQ ELTEGREKFK SRGVGELARL ALVIS.
Specifications
Specifications
| Antigen | TRIM26 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TRIM26 |
| Gene Accession No. | Q12899 |
| Gene Alias | Acid finger protein; AFP; AI462198; RING finger protein 95; RNF95; TRIM26; tripartite motif containing 26; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zfp173; Zfp1736; zinc finger protein 173; Znf173 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.