Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540836
Description
Peptide sequence: KKLQDQRQYI VAEFEQGHQF LREREEHLLE QLAKLEQELT EGREKFKSRG Sequence homology: Cow: 100%; Dog: 100%; Horse: 85%; Human: 100%; Mouse: 92%; Pig: 85%; Rabbit: 100%; Rat: 92%.
TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.
Specifications
TRIM26 | |
Polyclonal | |
Unconjugated | |
TRIM26 | |
Acid finger protein; AFP; AI462198; RING finger protein 95; RNF95; TRIM26; tripartite motif containing 26; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zfp173; Zfp1736; zinc finger protein 173; Znf173 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7726 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q12899 | |
TRIM26 | |
Synthetic peptide directed towards the middle region of human TRIM26. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction