Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560878 Supplier Invitrogen™ Supplier No. PA560878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESREHRPHTA VLMEKAAQPH REKILNHLST LRRDRDKIQG FQAKGEADIL AALKKLQDQR QYIVAEFEQG HQFLREREEH LLEQLAKLEQ ELTEGRE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM26
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM26
Gene Accession No. Q12899
Gene Alias Acid finger protein; AFP; AI462198; RING finger protein 95; RNF95; TRIM26; tripartite motif containing 26; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zfp173; Zfp1736; zinc finger protein 173; Znf173
Gene Symbols TRIM26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM26. Recombinant protein control fragment (Product #RP-99609).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.