Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220712
Description
TRIM27 Polyclonal specifically detects TRIM27 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRIM27 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
Rabbit | |
58 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P14373 | |
TRIM27 | |
Recombinant protein encompassing a sequence within the center region of human TRIM27. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
5987 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction