Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM27 Antibody (OTI4D4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274609AF405
Description
TRIM27 Monoclonal specifically detects TRIM27 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRIM27 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRIM27 | |
Full length recombinant protein of human TRIM27 (NP_006501) produced in E.coli. | |
0.1 mL | |
5987.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4D4 | |
Western Blot, Immunohistochemistry | |
Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction