Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM27 Antibody (PCRP-TRIM27-1B3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308579F
Description
TRIM27 Monoclonal antibody specifically detects TRIM27 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
TRIM27 | |
Monoclonal | |
FITC | |
Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
Recombinant full-length human TRIM27 protein (Uniprot: P14373) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-TRIM27-1B3 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
5987 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction