Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM27 Monoclonal Antibody (OTI3B12)
GREENER_CHOICE

Catalog No. PIMA526641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526641 Supplier Invitrogen™ Supplier No. MA526641
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM27 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nuclear matrix. It interacts with the enhancer of polycomb protein and represses gene transcription. It is also thought to be involved in the differentiation of male germ cells. Fusion of the N-terminus of this protein with the truncated C-terminus of the RET gene product has been shown to result in production of the ret transforming protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIM27
Gene Accession No. P14373
Gene Alias AW538890; Gm19403; ret finger protein; ret finger protein 1; RFP; RFP transforming protein; Rfp1; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; RNF76; Trim27; tripartite motif containing 27; tripartite motif protein 27; tripartite motif protein TRIM27; tripartite motif-containing 27; tripartite motif-containing protein 27; Zinc finger protein RFP
Gene Symbols TRIM27
Host Species Mouse
Immunogen Full length human recombinant protein of TRIM27 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5987
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.