Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727792 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7792 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7792 Supplier Proteintech Group Inc Supplier No. 122051AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM27, also designated Ret finger protein (RFP), is a member of the tripartite motif family, which is characterized by a conserved RING finger, a B-box and a coiled-coil domain (together designated RBCC). TRIM27 becomes oncogenic when its RBCC is fused with the tyrosine kinase domain of the RET protein.Normally, TRIM27 functions as a transcriptional repressor and associates with enhancer of polycomb (EPC1) and Mi-2beta, a main component of the nucleosome remodeling and deacetylase (NuRD) complex. TRIM27 is expressedat specific stages of spermatogenesis and in various tissues and can be localized to either the cytoplasm or nucleus depending on the cell type.

Specifications

Antigen TRIM27
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TRIM27
Gene Accession No. P14373, Q62158
Gene Alias Ret finger protein, RFP, RING finger protein 76, RNF76, TRIM27, tripartite motif containing 27, Zinc finger protein RFP
Gene Symbols Trim27
Host Species Rabbit
Immunogen TRIM27 Fusion Protein Ag2849
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19720, 291171, 5987
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.