Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM29 Antibody (SR2131), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32217850UL
Description
TRIM29 Monoclonal antibody specifically detects TRIM29 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRIM29 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
23650 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR2131 | |
Western Blot 1:500-1:2000 | |
Ataxia telangiectasia group D-associated protein, ATDC, ATDC ataxia-telangiectasia group D-associated protein, FLJ36085, tripartite motif containing 29, tripartite motif protein TRIM29, tripartite motif-containing 29, tripartite motif-containing protein 29 | |
A synthesized peptide derived from human TRIM29 (Uniprot #: Q14134) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction