Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM29 (Lung Squamous Cell Carcinoma Marker) Monoclonal Antibody (TRIM29), Abbomax

Catalog No. 502204441
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4441 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4441 Supplier Abbomax Inc Supplier No. 606370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: JAR cell lysate Cellular location: Cytoplasm.

TRIM29 (Lung Squamous Cell Carcinoma Marker) Monoclonal antibody specifically detects TRIM29 (Lung Squamous Cell Carcinoma Marker) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TRIM29 (Lung Squamous Cell Carcinoma Marker)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TRIM29
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Trim29
Gene Accession No. Q14134
Gene Alias 1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; ATDC; EGK_06995; FLJ36085; Trim29; tripartite motif containing 29; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; tripartite motif-containing protein 29
Host Species Mouse
Immunogen A recombinant protein of human TRIM29
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23650
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.