Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554131 Supplier Invitrogen™ Supplier No. PA554131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDDKNSNYFS MDSMEGKRSP YAGLQLGAAK KPPVTFAEKG ELRKSIFSES RKPTVSIMEP GETRRNSYPR ADTGLFSRSK SGSEEVLCDS CIGNKQKAVK SCLVCQASFC ELHL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM29
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trim29
Gene Accession No. Q14134
Gene Alias 1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; ATDC; EGK_06995; FLJ36085; Trim29; tripartite motif containing 29; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; tripartite motif-containing protein 29
Gene Symbols Trim29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM29. Recombinant protein control fragment (Product #RP-102417).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23650, 300656
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.