Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM3/BERP Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060400H
Description
TRIM3/BERP Polyclonal antibody specifically detects TRIM3/BERP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TRIM3/BERP | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BERPRING finger protein 97, Brain-expressed RING finger protein, HAC1, RING finger protein 22, RNF22brain expressed ring finger, RNF97FLJ16135, tripartite motif containing 3, tripartite motif protein TRIM3, tripartite motif-containing 3, tripartite motif-containing protein 3 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human tripartite motif-containing 3 (brain expressed ring finger protein) using the numbering given in entry NP_006449.2 (GeneID 10612). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10612 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction