Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM3/BERP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571747
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1747 100 μL
50-157-1748 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1747 Supplier Bethyl Laboratories, Inc Supplier No. IHC00243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Orangutan, Monkey, Gorill

The protein encoded by this gene is a member of the tripartite motif family, also called the ′RING-B-box-coiled-coil′ subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic filaments. It is similar to a rat protein which is a specific partner for the tail domain of myosin V, a class of myosins which are involved in the targeted transport of organelles. The rat protein can also interact with alpha-actinin-4. Thus it is suggested that this human protein may play a role in myosin V-mediated cargo transport. Alternatively spliced transcript variants encoding the same isoform have been identified.

Specifications

Antigen TRIM3/BERP
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TRIM3
Gene Accession No. O75382
Gene Alias TRIM3; BERP; RNF22; TRIM3; tripartite motif-containing 3; HAC1; RNF97; ring finger protein 22; brain expressed ring finger; tripartite motif protein TRIM3; FLJ16135
Gene Symbols TRIM3
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10612
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.