Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM3/BERP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TRIM3/BERP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TRIM3/BERP Polyclonal specifically detects TRIM3/BERP in Mouse samples. It is validated for Western Blot.Specifications
TRIM3/BERP | |
Polyclonal | |
Rabbit | |
NP_061368 | |
10612 | |
The immunogen for this antibody is Trim3. Peptide sequence FFISSLMEAMQQAPEGAHDPEDPHPLSAVAGRPLSCPNHEGKTMEFYCEA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
BERPRING finger protein 97, Brain-expressed RING finger protein, HAC1, RING finger protein 22, RNF22brain expressed ring finger, RNF97FLJ16135, tripartite motif containing 3, tripartite motif protein TRIM3, tripartite motif-containing 3, tripartite motif-containing protein 3 | |
TRIM3 | |
IgG | |
81 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title