Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540961 Supplier Invitrogen™ Supplier No. PA540961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CRESKDHKSH NVSLIEEAAQ NYQGQIQEQI QVLQQKEKET VQVKAQGVHR Sequence homology: Cow: 79%; Human: 100%; Mouse: 86%; Rabbit: 93%.

TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM31
Applications Immunohistochemistry, Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM31
Gene Accession No. Q8R0K2, Q9BZY9
Gene Alias BC026666; C6orf13; E3 ubiquitin-protein ligase TRIM31; HCG1; HCGI; RING-type E3 ubiquitin transferase TRIM31; RNF; Trim31; tripartite motif containing 31; tripartite motif protein 31; tripartite motif-containing 31; tripartite motif-containing protein 31
Gene Symbols TRIM31
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRIM31 (aa 117-166).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11074, 224762
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.