Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100793
Description
Antibody detects endogenous levels of total TRI31.
TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.
Specifications
TRIM31 | |
Polyclonal | |
Unconjugated | |
TRIM31 | |
BC026666; C6orf13; E3 ubiquitin-protein ligase TRIM31; HCG1; HCGI; RING-type E3 ubiquitin transferase TRIM31; RNF; Trim31; tripartite motif containing 31; tripartite motif protein 31; tripartite motif-containing 31; tripartite motif-containing protein 31 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11074 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BZY9 | |
TRIM31 | |
A synthesized peptide derived from human TRIM31(Accession Q9BZY9), corresponding to amino acid residues K318-G368. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction