Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5140269

This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ESKDHKSHNV SLIEEAAQNY QGQIQEQIQV LQQKEKETVQ VKAQGVHRVD VFTDQVEHEK QRILTEFELL HQVLEEEKNF LLS
TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.
Specifications
TRIM31 | |
Polyclonal | |
Unconjugated | |
TRIM31 | |
BC026666; C6orf13; E3 ubiquitin-protein ligase TRIM31; HCG1; HCGI; RING-type E3 ubiquitin transferase TRIM31; RNF; Trim31; tripartite motif containing 31; tripartite motif protein 31; tripartite motif-containing 31; tripartite motif-containing protein 31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZY9 | |
TRIM31 | |
Recombinant protein corresponding to Human TRIM31 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction