Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM31 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987132
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7132 Supplier Bioss Supplier No. BS6220R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.

Specifications

Antigen TRIM31
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRIM31
Gene Accession No. Q9BZY9
Gene Alias BC026666; C6orf13; E3 ubiquitin-protein ligase TRIM31; HCG1; HCGI; LOC100347988; RING-type E3 ubiquitin transferase TRIM31; RNF; TRIM31; tripartite motif containing 31; tripartite motif protein 31; tripartite motif-containing 31; Tripartite motif-containing protein 31
Gene Symbols TRIM31
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRIM31.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11074
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.