Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM32/BBS11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980604
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0604 Supplier Bioss Supplier No. BS0964R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.

Specifications

Antigen TRIM32/BBS11
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Trim32
Gene Accession No. Q13049
Gene Alias 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); BOS_9387; E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; Tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; Zinc finger protein HT2A; zinc-finger protein HT2A
Gene Symbols Trim32
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse TRIM32.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22954
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.