Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM32 Polyclonal Antibody

TRIM32 Polyclonal Antibody
Supplier: Thermo Scientific PA541535
Description
TRIM32 Polyclonal Antibody for Western Blot, IHC

Specifications
TRIM32 | |
Polyclonal | |
Unconjugated | |
Trim32 | |
1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A | |
Rabbit | |
Protein A | |
RUO | |
22954 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13049 | |
Trim32 | |
Synthetic peptide directed towards the C-terminal of human TRIM32 (aa 559-608). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction