Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562127 Supplier Invitrogen™ Supplier No. PA562127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLTPLSVAMN CQGLIGVTDS YDNSLKVYTL DGHCVACHRS QLSKPWGITA LPSGQFVVTD VEGGKLWCFT VDRGSGVVKY SCLCSAVRPK FVTCDA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM32
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trim32
Gene Accession No. Q13049
Gene Alias 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A
Gene Symbols Trim32
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM32. Recombinant protein control fragment (Product #RP-101224).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.