Learn More
Invitrogen™ TRIM32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110518
Description
Immunogen sequence: MAAAAASHLN LDALREVLEC PICMESFTEE QLRPKLLHCG HTICRQCLEK LLASSINGVR CPFCSKITRI TSLTQLTDNL TVLKIIDTAG LSEAVGLLMC RSCGRRLPRQ FCRSCGLVLC EPCREADHQP PGHCTLPVKE AAEERRRDFG EKLTRLRELM GELQRRKAAL EGVSKDLQAR YKAVLQEYGH EERRVQDELA RSRKFFTGSL AEVEKSNSQV VEEQSYLLNI AEVQAVSRCD YFLAKIKQAD VALLEETADE EEPELTASLP RELTLQDVEL.
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
Specifications
| TRIM32 | |
| Polyclonal | |
| Unconjugated | |
| Trim32 | |
| 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22954, 313264, 69807 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13049, Q8CH72 | |
| Trim32 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM32 (NP_036342.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.