Learn More
Invitrogen™ TRIM33 Monoclonal Antibody (16G9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1801
Description
MA1-801 detects the TIF1 gamma in human, rat and mouse cells. MA1-801 has been successfully used in Western blot, immunoprecipitation and immunofluorescence procedures. By Western blot, this antibody detects a 150 kDa protein. MA1-801 is specific for the gamma subunit of TIF1. The MA1-801 immunogen is recombinant human TIF gamma.
TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.
Specifications
TRIM33 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q99PP7, Q9UPN9 | |
TRIM33 | |
Recombinant protein: human TIF1 gamma. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
16G9 | |
Unconjugated | |
TRIM33 | |
8030451N04Rik; AI413936; E3 ubiquitin-protein ligase TRIM33; Ecto; Ectodermin homolog; Kiaa1113; Protein Rfg7; PTC7; RET-fused gene 7 protein; RFG7; RING-type E3 ubiquitin transferase TRIM33; TF1G; TIF 1-g; Tif1g; TIF1GAMMA; TIF1-gamma; TIFGAMMA; Transcription intermediary factor 1-gamma; transcriptional intermediary factor 1 gamma; Trim33; tripartite motif containing 33; tripartite motif protein 33; tripartite motif-containing 33; tripartite motif-containing protein 33 | |
Mouse | |
Protein G | |
RUO | |
365894, 51592, 94093 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.