Learn More
TRIM33/TIF1gamma Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301060AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The protein encoded by TIF1 gamma is thought to be a transcriptional corepressor. However, molecules that interact with this protein have not yet been identified. The protein is a member of the tripartite motif family. This motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Three alternatively spliced transcript variants for TIF1 gamma have been described, however, the full-length nature of one variant has not been determined.Specifications
TRIM33/TIF1gamma | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99PP7, Q9UPN9 | |
TRIM33 | |
Between 1077 and C-term | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
TRIM33 | |
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA, TRIM33, tripartite motif containing 33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51592, 94093 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.