Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567485 Supplier Invitrogen™ Supplier No. PA567485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRLEEEEKKTL DKFAEAEDEL VQQKQLVREL ISDVEC Highest antigen sequence identity to the following orthologs - mouse 62%, rat 62%.

TRIM34 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Expression of this gene is up-regulated by interferon. This gene is mappped to chromosome 11p15, where it resides within a TRIM gene cluster. Alternate splicing of this gene generates four transcript variants. Additionally, a read-through transcript transcribed from this gene and TRIM6 has been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM34
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM34
Gene Accession No. Q9BYJ4
Gene Alias IFP1; Interferon-responsive finger protein 1; RING finger protein 21; ring finger protein 21, interferon-responsive; RNF21; TRIM34; tripartite motif containing 34; tripartite motif-containing 34; tripartite motif-containing protein 34
Gene Symbols TRIM34
Host Species Rabbit
Immunogen Recombinant Human TRIM34. Recombinant protein control fragment (Product #RP-105672).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.