Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288473
Description
TRIM35 Polyclonal specifically detects TRIM35 in Human samples. It is validated for Western Blot.Specifications
TRIM35 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
HLS5Hemopoietic lineage switch protein 5, KIAA1098MGC17233, MAIR, tripartite motif containing 35, tripartite motif-containing 35, tripartite motif-containing protein 35 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of human TRIM35 | |
100 μL | |
Zinc Finger | |
23087 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction